siRNA Details
Gene Symbol: Elmod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253692.1 | NP_001240621.1 | 12 | 2191 | ||
NM_144917.5 | NP_659166.1 | 13 | 2222 | |||
GenBank mRNA | BC016193.1 | - | 11 | 1890 | 4487200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
