siRNA Details
Gene Symbol: Hnrnpul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144922.2 | NP_659171.1 | 13 | 2132 | ||
GenBank mRNA | AK087545.1 | - | 13 | 2103 | ||
BC021506.1 | - | 13 | 2156 | 5347099 | ||
BC027844.1 | - | 10 | 1425 | |||
BC078642.1 | - | 13 | 2056 | 6329630 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
