siRNA Details
Gene Symbol: Tnrc6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144925.3 | NP_659174.3 | 24 | 6822 | ||
GenBank mRNA | AK020685.1 | - | 1 | 26 | ||
AK044679.1 | - | 12 | 3206 | |||
AK050601.1 | - | 6 | 1702 | |||
AK138426.1 | - | 14 | 2872 | |||
AK142808.1 | - | 9 | 2191 | |||
AK142809.1 | - | 9 | 2191 | |||
AK147327.1 | - | 24 | 6527 | |||
BC005741.1 | - | 14 | 2319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
