siRNA Details
Gene Symbol: DIO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000793.5 | NP_000784.2 | 3, 4 | 616 | ||
NM_001242502.2 | NP_001229431.1 | 3, 4 | 627 | |||
NM_001242503.2 | NP_001229432.1 | 2, 3 | 519 | |||
NM_001324462.1 | NP_001311391.1 | 2, 3 | 632 | |||
NM_013989.4 | NP_054644.1 | 1, 2 | 385 | |||
GenBank mRNA | AB041844.1 | - | 3, 4 | 313 | ||
AF093774.1 | - | 1, 2 | 385 | |||
AK292210.1 | - | 3, 4 | 616 | |||
AK311603.1 | - | 2, 3 | 206 | |||
BC063118.1 | - | 1, 2 | 268 | |||
BC074882.2 | - | 1, 2 | 261 | 30915240 | ||
BC136514.1 | - | 1, 2 | 255 | 9020504 | ||
JK546617.1 | - | 2, 3 | 543 | |||
U53506.1 | - | 1, 2 | 364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
