siRNA Details
Gene Symbol: Ddx60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293783.1 | NP_001280712.1 | 32 | 4671 | ||
GenBank mRNA | BC013672.1 | - | 5 | 659 |

More Information
Other Species Targeted

Gene Ontology Categories:
