siRNA Details
Gene Symbol: Cd207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144943.3 | NP_659192.2 | 3 | 322 | ||
GenBank mRNA | AJ302711.1 | - | 3 | 322 | ||
AK142731.1 | - | 3 | 264 | |||
AY026050.1 | - | 3 | 264 | |||
AY078415.1 | - | 3 | 264 |

More Information
Other Species Targeted

Gene Ontology Categories:
