siRNA Details
Gene Symbol: Neto1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144946.4 | NP_659195.3 | 4 | 1169 | ||
GenBank mRNA | AF448840.1 | - | 4 | 502 | ||
AK032510.1 | - | 2 | 37 | |||
AK043710.1 | - | 4 | 505 | |||
AY138990.1 | - | 4 | 238 | |||
BC051145.1 | - | 4 | 529 | 6590345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
