siRNA Details
Gene Symbol: Slc25a28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145156.1 | NP_660138.1 | 4 | 999 | ||
GenBank mRNA | AF377993.1 | - | 3 | 468 | ||
AF377994.1 | - | 4 | 817 | |||
AK034173.1 | - | 1 | 1839 | |||
AK155044.1 | - | 2 | 3192 | |||
BC023172.1 | - | 2 | 510 | |||
BC025908.1 | - | 4 | 999 | 5043992 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
