siRNA Details
Gene Symbol: GART | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000819.4 | NP_000810.1 | 4 | 466 | ||
NM_001136005.1 | NP_001129477.1 | 4 | 642 | |||
NM_001136006.1 | NP_001129478.1 | 4 | 605 | |||
NM_175085.2 | NP_780294.1 | 4 | 466 | |||
XM_005260941.1 | XP_005260998.1 | 4 | 742 | |||
XM_006723989.1 | XP_006724052.1 | 4 | 684 | |||
XM_006723990.1 | XP_006724053.1 | 4 | 572 | |||
XM_011529526.1 | XP_011527828.1 | 4 | 651 | |||
GenBank mRNA | AB208785.1 | - | 4 | 532 | ||
AK292560.1 | - | 4 | 581 | |||
AK292897.1 | - | 4 | 419 | |||
BC038958.1 | - | 4 | 429 | |||
BC093641.1 | - | 3 | 385 | |||
BC101565.1 | - | 3 | 385 | |||
BC107712.1 | - | 4 | 420 | 6701741 | ||
BX648124.1 | - | 4 | 583 | |||
D32051.1 | - | 4 | 402 | |||
DB236855.1 | - | 4 | 466 | |||
X54199.1 | - | 4 | 409 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
