siRNA Details
Gene Symbol: BC089597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145424.2 | NP_663399.2 | 1 | 289 | ||
GenBank mRNA | AK144071.1 | - | 1 | 278 | ||
AY053571.1 | - | 1 | 209 | |||
AY053572.1 | - | 1 | 289 | |||
BC018263.1 | - | 1 | 285 | 4239862 | ||
BC089597.1 | - | 1 | 263 | 30120654 |
