siRNA Details
Gene Symbol: HSD3B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000862.2 | NP_000853.1 | 2 | 228 | ||
XM_011541314.2 | XP_011539616.1 | 2 | 269 | |||
XM_017001119.1 | XP_016856608.1 | 2 | 192 | |||
GenBank mRNA | AK291556.1 | - | 1 | 242 | ||
AL544564.3 | - | 2 | 228 | |||
AL550307.3 | - | 2 | 210 | |||
BC031999.1 | - | 1 | 170 | 4755300 | ||
M27137.1 | - | 1 | 86 | |||
M35493.1 | - | 1 | 113 | |||
S45679.1 | - | 2 | 214 | |||
X53321.1 | - | 1 | 113 | |||
X55997.1 | - | 1 | 83 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
