siRNA Details
Gene Symbol: Tmc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145439.2 | NP_663414.3 | 10 | 1396 | ||
GenBank mRNA | AK077671.1 | - | 8 | 1141 | ||
AK090179.1 | - | 10 | 1353 | |||
AY236497.1 | - | 10 | 1295 | |||
AY263158.1 | - | 10 | 1321 | |||
BC004840.1 | - | 10 | 1321 | 2648849 | ||
BC005510.1 | - | 3 | 366 | |||
BC006956.1 | - | 8 | 1106 | |||
BC013502.1 | - | 1 | 123 | 4223599 | ||
BC058195.1 | - | 10 | 1321 | 5356348 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
