siRNA Details
Gene Symbol: Mlh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029786.1 | - | 3, 4 | 2043 | ||
AK162464.1 | - | 2, 3 | 2118 | |||
BC003865.1 | - | 2, 3 | 245 | |||
BC072599.1 | - | 3, 4 | 3462 | |||
BC079861.1 | - | 3, 4 | 3449 | 6834767 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
