siRNA Details
Gene Symbol: Trim41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145377.2 | NP_663352.2 | 6 | 2127 | ||
GenBank mRNA | BC020156.1 | - | 4 | 332 | ||
BC041780.1 | - | 6 | 2089 |

More Information
Other Species Targeted

Gene Ontology Categories:
