siRNA Details
Gene Symbol: Tmc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145439.2 | NP_663414.3 | 10, 11 | 1453 | ||
GenBank mRNA | AK077671.1 | - | 8, 9 | 1198 | ||
AK090179.1 | - | 10, 11 | 1410 | |||
AY236497.1 | - | 10, 11 | 1352 | |||
AY263158.1 | - | 10, 11 | 1378 | |||
BC004840.1 | - | 10, 11 | 1378 | 2648849 | ||
BC005510.1 | - | 3, 4 | 423 | |||
BC006956.1 | - | 8, 9 | 1163 | |||
BC013502.1 | - | 1, 2 | 180 | 4223599 | ||
BC058195.1 | - | 10, 11 | 1378 | 5356348 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
