siRNA Details
Gene Symbol: Mlh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029786.1 | - | 4, 5 | 2148 | ||
AK132195.1 | - | 4, 5 | 3556 | |||
AK162464.1 | - | 3, 4 | 2223 | |||
BC003865.1 | - | 3, 4 | 350 | |||
BC072599.1 | - | 4, 5 | 3567 | |||
BC079861.1 | - | 4, 5 | 3554 | 6834767 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
