siRNA Details
Gene Symbol: Zbtb7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145356.3 | NP_663331.1 | 5 | 4104 | ||
GenBank mRNA | AK045522.1 | - | 6 | 3106 | ||
AK052470.1 | - | 1 | 1640 | |||
AK156372.1 | - | 3 | 3773 | |||
BC070424.1 | - | 5 | 4104 | 6837447 |
