siRNA Details
Gene Symbol: Alg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145362.2 | NP_663337.2 | 13 | 1727 | ||
GenBank mRNA | AK145871.1 | - | 13 | 1727 | ||
AK184910.1 | - | 1 | 387 | |||
BC011281.1 | - | 12 | 1627 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
