siRNA Details
Gene Symbol: Myo5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081322.1 | NP_001074791.1 | 36, 37 | 4529 | ||
GenBank mRNA | AK033571.1 | - | 12, 13 | 1334 | ||
BC003985.1 | - | 7, 8 | 985 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
