siRNA Details
Gene Symbol: Jmjd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114637.1 | NP_001108109.1 | 8 | 1118 | ||
GenBank mRNA | BC016255.1 | - | 8 | 1095 |
Gene Symbol: Gm28042 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB195274.1 | - | 8 | 1181 | ||
AB195275.1 | - | 8 | 1181 | |||
BC098210.1 | - | 2 | 342 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
