siRNA Details
Gene Symbol: Tmc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145439.2 | NP_663414.3 | 11, 12 | 1597 | ||
GenBank mRNA | AK077671.1 | - | 9, 10 | 1342 | ||
AK090179.1 | - | 11, 12 | 1554 | |||
AY236497.1 | - | 11, 12 | 1496 | |||
AY263158.1 | - | 11, 12 | 1522 | |||
BC004840.1 | - | 11, 12 | 1522 | 2648849 | ||
BC005510.1 | - | 4, 5 | 567 | |||
BC006956.1 | - | 9, 10 | 1307 | |||
BC013502.1 | - | 2, 3 | 324 | 4223599 | ||
BC058195.1 | - | 11, 12 | 1522 | 5356348 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
