siRNA Details
Gene Symbol: Mlh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029786.1 | - | 12 | 3076 | ||
AK132195.1 | - | 12 | 4484 | |||
AK162464.1 | - | 11 | 3151 | |||
AK163030.1 | - | 4 | 2526 | |||
BC003865.1 | - | 10 | 1118 | |||
BC072599.1 | - | 12 | 4495 | |||
BC079861.1 | - | 12 | 4482 | 6834767 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
