siRNA Details
Gene Symbol: Zswim6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145456.2 | NP_663431.2 | 14 | 3014 | ||
GenBank mRNA | AK037945.1 | - | 10 | 1609 | ||
AK122528.1 | - | 13 | 2282 | |||
BC021311.1 | - | 5 | 691 | |||
BC021521.1 | - | 5 | 894 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
