siRNA Details
Gene Symbol: Zfp521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145492.4 | NP_663467.1 | 3 | 404 | ||
GenBank mRNA | AK052655.1 | - | 3 | 307 | ||
AK131130.1 | - | 2 | 489 | |||
AY147406.1 | - | 3 | 307 | |||
BC021376.1 | - | 3 | 220 | 5038671 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
