siRNA Details
Gene Symbol: Cstf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145529.3 | NP_663504.1 | 2, 3 | 323 | ||
NM_177253.4 | NP_796227.1 | 2, 3 | 323 | |||
GenBank mRNA | AK028899.1 | - | 2, 3 | 268 | ||
AK159180.1 | - | 2, 3 | 126 | |||
BC003241.1 | - | 2, 3 | 284 | 3498424 | ||
BC058605.1 | - | 2, 3 | 275 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001077672, XM_008762067, BC127520.
Gene Ontology Categories:
