siRNA Details
Gene Symbol: Spg11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145531.2 | NP_663506.2 | 32, 33 | 6185 | ||
GenBank mRNA | AK031637.1 | - | 9, 10 | 2148 | ||
AK083468.1 | - | 1, 2 | 31 | |||
AK129453.1 | - | 19, 20 | 3619 | |||
AK147500.1 | - | 32, 33 | 6186 | |||
BC019404.1 | - | 3, 4 | 375 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
