siRNA Details
Gene Symbol: Zfp758 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145484.2 | NP_663459.2 | 4 | 773 | ||
GenBank mRNA | AK028301.1 | - | 3 | 657 | ||
BC021442.1 | - | 4 | 670 | 5038660 | ||
BC022203.1 | - | 4 | 750 | |||
BC037455.1 | - | 4 | 774 | |||
BC056632.1 | - | 3 | 631 | 5036441 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
