siRNA Details
Gene Symbol: Pprc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081214.1 | NP_001074683.1 | 5 | 3273 | ||
GenBank mRNA | AK044522.1 | - | 5 | 3220 | ||
AK122322.1 | - | 5 | 3231 | |||
AK147947.1 | - | 5 | 3254 | |||
BC013720.1 | - | 1 | 640 | |||
BC066048.1 | - | 5 | 3227 | 6825158 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
