siRNA Details
Gene Symbol: Ormdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145517.4 | NP_663492.3 | 4 | 666 | ||
GenBank mRNA | AK050385.1 | - | 4 | 651 | ||
BC012315.1 | - | 4 | 621 | 3981845 | ||
BC023695.1 | - | 4 | 646 | 5344622 | ||
BC023903.1 | - | 4 | 609 | |||
BC025572.1 | - | 4 | 642 | 5323819 | ||
EL956043.1 | - | 1 | 164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
