siRNA Details
Gene Symbol: Zswim6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145456.2 | NP_663431.2 | 14 | 3603 | ||
GenBank mRNA | AK037945.1 | - | 10 | 2198 | ||
BC021311.1 | - | 5 | 1280 | |||
BC021521.1 | - | 5 | 1483 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
