siRNA Details
Gene Symbol: Zfp758 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145484.2 | NP_663459.2 | 4 | 1319 | ||
GenBank mRNA | AK028301.1 | - | 3 | 1203 | ||
BC021442.1 | - | 4 | 1216 | 5038660 | ||
BC022203.1 | - | 4 | 1296 | |||
BC037455.1 | - | 4 | 1320 | |||
BC056632.1 | - | 3 | 1177 | 5036441 |

More Information
Other Species Targeted

Gene Ontology Categories:
