siRNA Details
Gene Symbol: Cstf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145529.3 | NP_663504.1 | 21 | 2456 | ||
GenBank mRNA | AK159180.1 | - | 21 | 2259 | ||
AK160306.1 | - | 7 | 992 | |||
BC003241.1 | - | 21 | 2417 | 3498424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
