siRNA Details
Gene Symbol: Spg11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145531.2 | NP_663506.2 | 40 | 7368 | ||
GenBank mRNA | AK031637.1 | - | 17 | 3334 | ||
AK083468.1 | - | 9 | 1214 | |||
AK129453.1 | - | 27 | 4802 | |||
AK147500.1 | - | 40 | 7369 | |||
BC019404.1 | - | 11 | 1558 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
