siRNA Details
Gene Symbol: Tmem161a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293797.1 | NP_001280726.1 | 4 | 297 | ||
NM_145597.4 | NP_663572.1 | 5 | 401 | |||
GenBank mRNA | AK049979.1 | - | 5 | 400 | ||
AK082758.1 | - | 5 | 397 | |||
AK082916.1 | - | 4 | 298 | |||
AK088569.1 | - | 4 | 287 | |||
AK147962.1 | - | 5 | 394 | |||
BC021367.1 | - | 5 | 401 | 5030434 |
