siRNA Details
Gene Symbol: Herc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145617.3 | NP_663592.3 | 2 | 275 | ||
GenBank mRNA | AK047003.1 | - | 2 | 276 | ||
AK083823.1 | - | 2 | 201 | |||
BC004027.1 | - | 2 | 194 |

More Information
Other Species Targeted

Gene Ontology Categories:
