siRNA Details
Gene Symbol: Sbno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081203.1 | NP_001074672.1 | 4 | 526 | ||
GenBank mRNA | AB188811.1 | - | 3 | 455 | ||
AK077133.1 | - | 4 | 532 | |||
AK164542.1 | - | 4 | 605 | |||
AK165852.1 | - | 3 | 465 | |||
BC021875.1 | - | 3 | 472 | 4212192 | ||
BC106156.1 | - | 4 | 585 | |||
BC138418.1 | - | 4 | 583 | 8861438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
