siRNA Details
Gene Symbol: Cd300lh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199201.1 | NP_954671.1 | 2 | 141 | ||
GenBank mRNA | AY457049.1 | - | 2 | 141 | ||
BC119352.1 | - | 2 | 176 | 8734101 | ||
BC120623.1 | - | 176 | 40129546 |
Gene Symbol: Cd300lf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169153.1 | NP_001162624.1 | 2 | 255 | ||
NM_145634.3 | NP_663609.2 | 2 | 255 | |||
GenBank mRNA | AB292061.1 | - | 2 | 144 | ||
AF251703.1 | - | 2 | 209 | |||
AK089313.1 | - | 2 | 255 | |||
AK089465.1 | - | 2 | 254 | |||
AK154273.1 | - | 2 | 188 | |||
AK155367.1 | - | 2 | 253 | |||
AY214460.1 | - | 2 | 207 | |||
AY457047.1 | - | 2 | 144 | |||
BC057864.1 | - | 2 | 220 | 5318856 | ||
JX073136.1 | - | 2 | 230 |
Gene Symbol: Gm11710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101656.2 | NP_001095126.1 | 176 |
Gene Symbol: Gm11711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101657.2 | NP_001095127.1 | 2 | 176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
