siRNA Details
Gene Symbol: Tardbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003898.3 | NP_001003898.1 | 3 | 692 | ||
NM_001003899.2 | NP_001003899.1 | 3 | 692 | |||
NM_001008545.2 | NP_001008545.1 | 3 | 692 | |||
NM_001008546.2 | NP_001008546.1 | 3 | 692 | |||
NM_145556.4 | NP_663531.1 | 3 | 692 | |||
GenBank mRNA | AK032199.1 | - | 3 | 462 | ||
AK045495.1 | - | 3 | 462 | |||
AK076208.1 | - | 3 | 461 | |||
AK083312.1 | - | 3 | 466 | |||
AK133207.1 | - | 3 | 469 | |||
AK146002.1 | - | 3 | 462 | |||
AK146997.1 | - | 4 | 371 | |||
AK153803.1 | - | 3 | 466 | |||
AY319513.1 | - | 2 | 362 | |||
AY319514.1 | - | 2 | 362 | |||
AY319515.1 | - | 2 | 362 | |||
AY319516.1 | - | 2 | 362 | |||
BB614725.1 | - | 3 | 715 | |||
BC012873.1 | - | 3 | 450 | 4016437 | ||
BC025544.1 | - | 3 | 463 | 5321007 | ||
BC027105.1 | - | 3 | 454 | 5359082 | ||
BC027772.1 | - | 3 | 452 | 5346061 | ||
BC029728.1 | - | 3 | 453 | 4950263 | ||
BC031126.1 | - | 3 | 454 | 4977114 | ||
BC033475.1 | - | 3 | 450 | 4953334 | ||
GQ428775.1 | - | 2 | 362 | |||
GQ428776.1 | - | 2 | 362 | |||
GQ428777.1 | - | 2 | 362 | |||
GU250735.1 | - | 2 | 362 |
