siRNA Details
Gene Symbol: Zfp932 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145563.2 | NP_663538.2 | 4 | 1808 | ||
GenBank mRNA | AK032220.1 | - | 4 | 1349 | ||
BC012405.1 | - | 4 | 1794 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
