siRNA Details
Gene Symbol: Ccdc136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201378.1 | NP_001188307.1 | 12 | 1898 | ||
NM_145574.3 | NP_663549.3 | 12 | 1898 | |||
GenBank mRNA | AK160245.1 | - | 5 | 625 | ||
BC006583.1 | - | 10 | 1486 | |||
EU029664.1 | - | 12 | 1898 | |||
EU029665.1 | - | 12 | 1898 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
