siRNA Details
Gene Symbol: Ttc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145607.3 | NP_663582.2 | 13 | 1503 | ||
GenBank mRNA | AK144842.1 | - | 13 | 1504 | ||
AK148102.1 | - | 15 | 1733 | |||
AK160894.1 | - | 15 | 1741 | |||
BC017545.1 | - | 7 | 717 | |||
BC072577.1 | - | 13 | 1496 | |||
BC079851.1 | - | 14 | 1558 | 6401930 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
