siRNA Details
Gene Symbol: Kank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145611.4 | NP_663586.3 | 3 | 820 | ||
GenBank mRNA | AK031324.1 | - | 3 | 811 | ||
AK049288.1 | - | 3 | 821 | |||
BC006647.1 | - | 3 | 808 | |||
BC010245.1 | - | 3 | 777 | 3710765 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
