siRNA Details
Gene Symbol: Herc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145617.3 | NP_663592.3 | 2 | 302 | ||
GenBank mRNA | AK047003.1 | - | 2 | 303 | ||
AK083823.1 | - | 2 | 228 | |||
BC004027.1 | - | 2 | 221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
