siRNA Details
Gene Symbol: Rrp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145620.4 | NP_663595.1 | 3 | 229 | ||
GenBank mRNA | AF368232.1 | - | 3 | 212 | ||
AK076113.1 | - | 3 | 231 |

More Information
Other Species Targeted

Gene Ontology Categories:
