siRNA Details
Gene Symbol: Obox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145708.2 | NP_663754.2 | 2 | 408 | ||
GenBank mRNA | BC094912.1 | - | 2 | 288 | ||
CA556780.1 | - | 2 | 308 | |||
DV663188.1 | - | 2 | 407 |
Gene Symbol: Obox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027802.2 | NP_082078.1 | 2 | 311 | ||
GenBank mRNA | AK018362.1 | - | 2 | 308 | ||
AK136061.1 | - | 2 | 301 | |||
BC141323.1 | - | 271 | 9055935 | |||
BC141324.1 | - | 271 | 9055936 |
Gene Symbol: Obox7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038676.1 | NP_001033765.1 | 2 | 325 | ||
GenBank mRNA | BC099488.1 | - | 2 | 325 | 30939375 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
