siRNA Details
Gene Symbol: Obox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027802.2 | NP_082078.1 | 4 | 957 | ||
GenBank mRNA | AK018362.1 | - | 4 | 954 | ||
AK136061.1 | - | 4 | 947 | |||
BC141323.1 | - | 917 | 9055935 | |||
BC141324.1 | - | 917 | 9055936 |
Gene Symbol: Obox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145708.2 | NP_663754.2 | 4 | 1049 | ||
GenBank mRNA | BC094912.1 | - | 4 | 929 | ||
DV647958.1 | - | 1 | 437 |
Gene Symbol: Obox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145709.2 | NP_663755.2 | 4 | 1024 | ||
GenBank mRNA | BB701159.1 | - | 1 | 193 | ||
BC052664.1 | - | 4 | 1028 | 30053785 | ||
DV653414.1 | - | 2 | 589 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
