siRNA Details
Gene Symbol: Slc2a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012363.2 | NP_001012363.2 | 12 | 2952 | ||
NM_001102414.1 | NP_001095884.1 | 12 | 3151 | |||
NM_001102415.1 | NP_001095885.1 | 10 | 2830 | |||
NM_145559.2 | NP_663534.1 | 11 | 2825 | |||
GenBank mRNA | AK033725.1 | - | 5 | 2022 | ||
AK037093.1 | - | 11 | 2816 | |||
AK156740.1 | - | 12 | 3055 | |||
AK172234.1 | - | 12 | 3055 | |||
BC006076.1 | - | 11 | 2825 | 3593581 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
