siRNA Details
Gene Symbol: Ttc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145607.3 | NP_663582.2 | 14, 15 | 1683 | ||
GenBank mRNA | AK144842.1 | - | 14, 15 | 1684 | ||
AK148102.1 | - | 16, 17 | 1913 | |||
AK160894.1 | - | 16, 17 | 1920 | |||
BC017545.1 | - | 8, 9 | 894 | |||
BC072577.1 | - | 14, 15 | 1673 | |||
BC079851.1 | - | 15, 16 | 1738 | 6401930 |

More Information
Other Species Targeted

Gene Ontology Categories:
