siRNA Details
Gene Symbol: Sbno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081203.1 | NP_001074672.1 | 9 | 1164 | ||
GenBank mRNA | AB188811.1 | - | 8 | 1093 | ||
AK077133.1 | - | 9 | 1170 | |||
AK164542.1 | - | 8 | 2604 | |||
AK165852.1 | - | 8 | 1103 | |||
BC021875.1 | - | 8 | 1110 | 4212192 | ||
BC023706.1 | - | 5 | 532 | |||
BC106156.1 | - | 9 | 1223 | |||
BC138418.1 | - | 9 | 1221 | 8861438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
