siRNA Details
Gene Symbol: Obox7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038676.1 | NP_001033765.1 | 2 | 299 | ||
GenBank mRNA | BC099488.1 | - | 2 | 299 | 30939375 |
Gene Symbol: Obox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145707.3 | NP_663753.3 | 3 | 347 | ||
GenBank mRNA | AK139750.1 | - | 3 | 348 | ||
AY094981.1 | - | 737 | ||||
BC119564.1 | - | 1 | 609 |
Gene Symbol: Obox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145709.2 | NP_663755.2 | 2 | 340 | ||
GenBank mRNA | BC052664.1 | - | 2 | 344 | 30053785 | |
CJ052752.1 | - | 2 | 342 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
